missing translation for 'onlineSavingsMsg'
Learn More

Corning™ XT Cooling Core for CoolBox™ XT Workstation

Allows for continuous uninterupted cooling. Corning™ XT Cooling Core for CoolBox™ XT Workstation provides cooling source workstation when thermo-conductive CoolRack or CoolSink tube and plate module is placed on top.

Brand:  Corning™ 432081

Code : NEW

Additional Details : Weight : 1.00000kg

Product Code. 15562851

  • 183.00 € / Each


Estimated Shipment
23-03-2020
Add to basket

Description

Description

  • Provides over 16 hours of 0.5 to 4°C cooling with lid on and over 10 hours with lid off
  • In XT Starter cooling system, XT Cooling Core provides over 4 hours of 2°C cooling to all samples
  • Pre-freeze in a −20° freezer until frozen, then place in CoolBox XT base when ready to work
Specifications

Specifications

XT Cooling Core
Maintaining sample temperature
16.2 x 12.1 x 6.1 cm (6.3 x 4.7 x 2.1 in.)
1
CoolBox XT, CoolBox 2XT, or XT Starter Systems
Videos
Documents