missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plastic Radiation Hazard Trays
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
70.40 € - 153.00 €
Specifications
Color | Yellow |
---|---|
Material | Plastic |
Type | Radiation Hazard Tray |
For Use With | Safe containment of spillages, lab bench protection |
Includes | Liner |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12357247
|
Fisherbrand™
|
113 x 54cm |
153.00 €
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
12307247
|
Fisherbrand™
|
46 x 26cm |
70.40 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12327247
|
Fisherbrand™
|
57 x 54cm |
79.75 €
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
Description
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Yellow | |
Radiation Hazard Tray | |
Liner | |
Rectangular |
Plastic | |
Safe containment of spillages, lab bench protection | |
1 Each |