missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Cables and DO/Temp/Conductivity Probes for 556 Multi-Probe System

Handheld multiprobe meter

Brand:  YSI™ 556320

Code : 03

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11950115

  • 2940.00 € / Each


Estimated Shipment: 28-07-2021
Add to basket

Description

Description

DO/Temperature/Conductivity Probe and Cable. Required to measure dissolved oxygen, temperature, conductivity.

Specifications

Specifications

Measures DO, temperature, conductivity
DO/Temperature/Conductivity
64 ft.
Temperature/conductivity sensor
YSI model 556 handheld multi probe meter
1 Year
20 m
Videos
SDS
Documents
Certificates