missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System
Measure pH and pH & ORP/Redox quickly and easily
Brand: YSI™ 005564
Code : A0
Additional Details : Weight : 0.80000kg
Description
- User-replaceble sensor allows for quick sensor replacements if needed
- No tools necessary
Specifications
pH Probe | |
Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications | |
YSI model 556 handheld multi probe meter | |
Storage bottle with storage solution |