missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ pH Probes for Pro Series Portable Meters
Choose from lab-grade and field models.
189.00 €
Specifications
For Use With (Application) | pH |
---|---|
For Use With (Equipment) | Pro series meter |
Description
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty. Measure pH quickly and easily with the 1001 sensor. The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.
For use with Pro Series Portable Meters.Order Info
Lab-grade models come with cable and connector. Field model requires cable/connector assembly 15-177-466.
Specifications
pH | |
Pro series meter |