missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DREF Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0846
Code : NEW
Additional Details : Weight : 0.01000kg
Description
ZBED1 Synthetic Peptide, PEP-0846, from Invitrogen.
This peptide corresponds to 15 amino acids near the center of human ZBED1.PEP-0846 can be used as a blocking peptide with polyclonal antibody PA5-20732.Specifications
9189 | |
purified | |
Human DREF | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O96006 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
ZBED1 | |
0.02% sodium azide | |
50 μg | |
PBS |