Learn More
Fisher Bioreagents Zeocin used as a selective agent for the Sh ble gene from Streptomyces verticullus TM-Zeocin is a tradement of Cayla - store at 0-5°C Fisher BioReagents
Brand:  Fisher Bioreagents ZO186-1G
Zeocin used as a selective agent for the Sh ble gene from Streptomyces verticullus TM-Zeocin is a tradement of Cayla - store at 0-5°C Fisher BioReagents
Code : 32
Additional Details : Weight : 0.00100kg
Product Code. 12821610