missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3593547
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
5970-45-6
C4H6O4Zn·2H2O
219.5
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
5970-45-6
C4H6O4Zn·2H2O
219.5
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 237.0°C
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CH3CO2)2Zn·2H2O
Merck Index 15, 10327
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 219.5
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.84
Density 1.8400g/mL
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'