missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, ≥97%, (Crystalline/ACS Reagent Grade), MP Biomedicals
36.42 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Use promo code "23257" to get your promotional price.
Click to view available options
Quantity:
100 g
Specifications
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | ≥97% |
Formula Weight | 136.28 |
Grade | ACS Reagent |
Chemical Name or Material | Zinc Chloride |
Safety and Handling
Product Identifier
- Zinc Chloride, ACS
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : zinc chloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'