missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2656, 50% in H2O, colloidal dispersion with nonionic dispersant and rheology modifier

Brand:  Alfa Aesar™ 047222.22

Code : NEW

Additional Details : CAS Number : 1314-13-2 Weight : 0.10000kg Transport : UN number : 3082 Chem class : 9 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 15475075

  • 36.40 € / 100g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]

Specifications

Zinc oxide
70nm APS for dry (typical)
NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier
OZn
UN3082
50% in H2O
O=[Zn]
81.379
CHEBI:36560
Powder
Odorless
100g
1314-13-2
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
50% in H2O
Videos
SDS