missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulphate, Reagecon™
Click to view available options
Quantity:
1 L
Specifications
Quantity | 1 L |
Concentration or Composition (by Analyte or Components) | 0.1mol/L |
Safety and Handling
Product Identifier
- ZINC SULPHATE 0.1M (0.1N)
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : zinc sulphate (hydrous) (mono-, hexa-andhepta hydrate)
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'