Learn More
Merck ZipTip™ Pipet Tips
Sample preparation and precise liquid handling in one device
Brand: Merck ZTC18M008
Code : 72
Additional Details : Weight : 0.05000kg
Description
Sample preparation and precise liquid handling in one device
A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.
- Concentrate and purify 1–100μL biological samples
- Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques
Specifications
With Micro Bed of C18 Media | |
1.2 in. | |
Specialty, Media-Fixed | |
Universal Fit | |
Polypropylene |
10 μL | |
3cm | |
8/Pk. | |
ZipTip |