missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ZM-336372
A potent and specific inhibitor of the protein kinase c-Raf
46.35 € - 269.00 €
Specifications
Description | Off-white |
---|---|
Content And Storage | -30°C to -10°C |
For Use With (Application) | A potent and specific inhibitor of the protein kinase c-Raf |
CAS | 208260-29-1 |
Molecular Formula | C23H23N3O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447049
|
Thermo Scientific™
J65730.#0 |
5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15457049
|
Thermo Scientific™
J65730.M^ |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Off-white | |
A potent and specific inhibitor of the protein kinase c-Raf | |
C23H23N3O3 | |
N-[5-(3-Dimethylaminobenzamido)-2-methylphenyl]-4-hydroxybenzamide; | |
ZM-336372 |
-30°C to -10°C | |
208260-29-1 | |
389.45 | |
Light Sensitive | |
Solid |