Learn More
Emma Safety Footwear Zolder Safety Shoes
Water-repellant oiled Nubuck uppers with internal steel toe protection and an exceptional 'sticky' grip, even on wet and greasy surfaces.
Brand:  Emma Safety Footwear MM50884639
Additional Details : Weight : 1.64000kg
 View more versions of this product
Product Code. 15988641
  • 151.58 € / 1 pair
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo/HRO
  • Oiled Nubuck
  • Heat Resistant Outsole
Specifications
Specifications
EN ISO 20345:2011
Zolder
Oiled Nubuck
Brown
Full grain Nappa leather / Split leather
39