Learn More
Emma Safety Footwear Zolder XD Safety Shoes
Water-repellant oiled Nubuck uppers with internal steel toe protection and an exceptional 'sticky' grip, even on wet and greasy surfaces.
Brand:  Emma Safety Footwear MM51886645
Additional Details : Weight : 1.91000kg
 View more versions of this product
Product Code. 15918661
  • 151.58 € / 1 pair
Estimated Shipment: 07-10-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo/HRO
  • Oiled Nubuck
  • Heat Resistant Outsole
Specifications
Specifications
Zolder XD
Oiled Nubuck
45