CAS RN 100080-82-8
CAS RN 100080-82-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichloroethylenebis(indenyl)zirconium(IV), racemic mixture, Thermo Scientific™
CAS: 100080-82-8 MDL Number: MFCD01314544 Synonym: Ethylenebis(indenyl)zirconium(IV) dichloride