missing translation for 'onlineSavingsMsg'
Learn More

Dichloroethylenebis(indenyl)zirconium(IV), racemic mixture, Thermo Scientific™

Product Code. 11398408
Click to view available options
Quantity:
0.5 g
2.5 g
Unit Size:
0.5g
2.5g
This item is not returnable. View return policy

Product Code. 11398408

Brand: Thermo Scientific Alfa Aesar 040449.02

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Useful for the preparation of metallocene type catalysts

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100080-82-8
MDL Number MFCD01314544
Synonym Ethylenebis(indenyl)zirconium(IV) dichloride

Specifications

Quantity 0.5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 418.48
Physical Form Powder
Chemical Name or Material Dichloroethylenebis(indenyl)zirconium(IV), Racemic mixture

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.