accessibility menu, dialog, popup

UserName

HNNHNHHNHClHClHClHCl

CAS RN 10045-25-7

IUPAC Name: 1,4,7,10-tetraazacyclododecane tetrahydrochloride
Synonyms: cyclen tetrahydrochloride
Molecular Weight (g/mol): 318.11
Molecular Formula: C8H24Cl4N4
InChi Key: SBWLCGZEBQGYRP-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.C1CNCCNCCNCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results