missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclododecane tetrahydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11465926
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465926 0.25 g 0.25g
11445926 1 g 1g
11455926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465926 Supplier Thermo Scientific Alfa Aesar Supplier No. B22268.01
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10045-25-7
Molecular Formula C8H24Cl4N4
Molecular Weight (g/mol) 318.108
MDL Number MFCD00068484
InChI Key SBWLCGZEBQGYRP-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclododecane tetrahydrochloride, cyclen tetrahydrochloride, cyclen-4hcl, unii-51830y7tix, tetraaza-12-crown-4 tetrahydrochloride, 1,4,7,10-tetraazacyclododecane, tetrahydrochloride, 1,4,7,10-tetraazacyclododecane, hydrochloride 1:4, 1,4,7,10-tetraazacyclododecanetetrahydrochloride, cyclen tetrahydrochloride mi, 1,4,7,10-tetraazacyclododecane, chloride, chloride, chloride, chloride
PubChem CID 11652630
IUPAC Name 1,4,7,10-tetrazacyclododecane;tetrahydrochloride
SMILES C1CNCCNCCNCCN1.Cl.Cl.Cl.Cl

Specifications

Melting Point >290°C
Odor Odorless
Quantity 0.25 g
UN Number UN3077
Sensitivity Hygroscopic
Formula Weight 318.12
Percent Purity 98%
Chemical Name or Material 1,4,7,10-Tetraazacyclododecane tetrahydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.