accessibility menu, dialog, popup

UserName

CAS RN 1008119-70-7

IUPAC Name: [3-fluoro-4-(morpholine-4-carbonyl)phenyl]boronic acid
Synonyms: 3-fluoro-4-(morpholine-4-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 253.04
Molecular Formula: C11H13BFNO4
InChi Key: PCOQCLUJTVPMSY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=C(C=C1)C(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results