accessibility menu, dialog, popup

UserName

CH3OFH2NF

CAS RN 1017778-10-7

IUPAC Name: 3-ethoxy-2,4-difluoroaniline
Synonyms: 3-ethoxy-2,4-difluoroaniline
Molecular Weight (g/mol): 173.16
Molecular Formula: C8H9F2NO
InChi Key: SVBJINNDWHYYHC-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=CC(N)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results