accessibility menu, dialog, popup

UserName

CAS RN 1017778-72-1

CH3OClOHOF

CAS RN 1017778-72-1

IUPAC Name: 3-chloro-4-ethoxy-5-fluorobenzoic acid
Synonyms: 3-chloro-4-ethoxy-5-fluorobenzoic acid
Molecular Weight (g/mol): 218.61
Molecular Formula: C9H8ClFO3
InChi Key: UZGWOMNUSFRIRW-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(C=C1Cl)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results