accessibility menu, dialog, popup

UserName

CAS RN 103-55-9

H3CNCH3NH

CAS RN 103-55-9

IUPAC Name: benzyl[2-(dimethylamino)ethyl]amine
Synonyms: benzyl[2-(dimethylamino)ethyl]amine
Molecular Weight (g/mol): 178.28
Molecular Formula: C11H18N2
InChi Key: LLSJAFHDYCTFCM-UHFFFAOYSA-N
SMILES: CN(C)CCNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results