accessibility menu, dialog, popup

UserName

FNH2H2NFHClHCl

CAS RN 1055301-10-4

IUPAC Name: 1,2-bis(4-fluorophenyl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(4-fluorophenyl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 321.19
Molecular Formula: C14H16Cl2F2N2
InChi Key: ILMQMKZMXREKBI-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=CC=C(F)C=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results