missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-1,2-Bis(4-fluorophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™

Product Code. 11445770
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445770 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445770 Supplier Thermo Scientific Alfa Aesar Supplier No. H25976.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1055301-10-4
Molecular Formula C14H16Cl2F2N2
Molecular Weight (g/mol) 321.193
MDL Number MFCD09265298
InChI Key ILMQMKZMXREKBI-KFWOVWKUSA-N
Synonym r,r-bis-4-fluorophenyl ethylenediamine dihydrochloride, r,r-1,2-bis 4-fluorophenyl-1,2-ethanediamine dihydrochloride, 1r,2r-1,2-bis 4-fluorophenyl ethane-1,2-diamine dihydrochloride, r,r-1,2-bis 4-fluorophenyl-1,2-ethanediamine dihydrochloride, ee
PubChem CID 45925687
IUPAC Name (1R,2R)-1,2-bis(4-fluorophenyl)ethane-1,2-diamine;dihydrochloride
SMILES C1=CC(=CC=C1C(C(C2=CC=C(C=C2)F)N)N)F.Cl.Cl

Specifications

Melting Point 200°C
Quantity 1 g
Optical Rotation 41° (c=1 in Methanol)
Formula Weight 321.21
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (R,R)-1,2-Bis(4-fluorophenyl)-1,2-ethanediamine dihydrochloride
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.