accessibility menu, dialog, popup

UserName

NSBr

CAS RN 106086-78-6

IUPAC Name: 2-(bromomethyl)-1,3-benzothiazole
Synonyms: 2-(bromomethyl)-1,3-benzothiazole
Molecular Weight (g/mol): 228.11
Molecular Formula: C8H6BrNS
InChi Key: WFLCAOGKZQTOIG-UHFFFAOYSA-N
SMILES: BrCC1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)