missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)benzothiazole, 98+%

Product Code. 15453397
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453397 250 mg 250mg
11342109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453397 Supplier Thermo Scientific Alfa Aesar Supplier No. H26120.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 106086-78-6 | C8H6BrNS | 228.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106086-78-6
Molecular Formula C8H6BrNS
Molecular Weight (g/mol) 228.107
MDL Number MFCD00461195
InChI Key WFLCAOGKZQTOIG-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole
PubChem CID 2776258
IUPAC Name 2-(bromomethyl)-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)CBr

Specifications

Melting Point 47°C to 49°C
Quantity 250 mg
UN Number UN3261
Formula Weight 228.11
Percent Purity ≥98%
Chemical Name or Material 2-(Bromomethyl)benzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.