accessibility menu, dialog, popup

UserName

CAS RN 1071466-57-3

IUPAC Name: 4-{2-[4-(acetyloxy)phenyl]propan-2-yl}phenyl propanoate
Synonyms: 4-{2-[4-(acetyloxy)phenyl]propan-2-yl}phenyl propanoate
Molecular Weight (g/mol): 326.39
Molecular Formula: C20H22O4
InChi Key: VWMUCEDXVAQLNS-UHFFFAOYSA-N
SMILES: CCC(=O)OC1=CC=C(C=C1)C(C)(C)C1=CC=C(OC(C)=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results