missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A acetate propionate, epoxy curative, EC-326, Thermo Scientific™

Product Code. 15447215
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447215 100 g 100g
15437215 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447215 Supplier Thermo Scientific Alfa Aesar Supplier No. 046441.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Recommended for use as an epoxy curative to improve toughness, hydrophobicity, and hydrolytic stability
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071466-57-3
Molecular Formula C20H22O4
Molecular Weight (g/mol) 326.392
MDL Number MFCD22380701
InChI Key VWMUCEDXVAQLNS-UHFFFAOYSA-N
Synonym bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate
PubChem CID 59317407
IUPAC Name [4-[2-(4-acetyloxyphenyl)propan-2-yl]phenyl] propanoate
SMILES CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C

Specifications

Color White to Yellow
Physical Form White to yellow solid
Quantity 100 g
Formula Weight 326.39
Chemical Name or Material Bisphenol A acetate propionate, Epoxy curative, EC-326
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.