accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COHN

CAS RN 1073353-57-7

IUPAC Name: N-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: N-benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 337.23
Molecular Formula: C20H24BNO3
InChi Key: VAFYISMQZLEYKR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results