CAS RN 107381-32-8
CAS RN 107381-32-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Propafenone, TRC
CAS: 107381-32-8 Molecular Formula: C21 H27 N O3 Molecular Weight (g/mol): 341.44 Synonym: 1-Propanone, 1-[2-[(2S)-2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-,1-Propanone, 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, (S)-,1-[2-[(2S)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-1-propanone,(-)-(S)-Propafenone,(-)-Propafenone,(S)-Propafenone IUPAC Name: 1-[2-[(2S)-2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one SMILES: CCCNC[C@H](O)COc1ccccc1C(=O)CCc2ccccc2