missing translation for 'onlineSavingsMsg'
Learn More

(S)-Propafenone, TRC

Product Code. 30419762
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419762 50 mg 50mg
30412510 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30419762 Supplier Toronto Research Chemicals Supplier No. TRCP75749650MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107381-32-8
Molecular Formula C21 H27 N O3
Molecular Weight (g/mol) 341.44
Synonym 1-Propanone, 1-[2-[(2S)-2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, 1-Propanone, 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-, (S)-, 1-[2-[(2S)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-1-propanone, (-)-(S)-Propafenone, (-)-Propafenone, (S)-Propafenone
IUPAC Name 1-[2-[(2S)-2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
SMILES CCCNC[C@H](O)COc1ccccc1C(=O)CCc2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H27NO3/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17/h3-11,18,22-23H,2,12-16H2,1H3/t18-/m0/s1
Formula Weight 341.1991
Chemical Name or Material (S)-Propafenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.