accessibility menu, dialog, popup

UserName

CH3OONONCH3CH3NONFPFFFFF(E/Z)

CAS RN 1075198-30-9

IUPAC Name: ethyl cyano({[(dimethyliminiumyl)(morpholin-4-yl)methoxy]imino})formate; hexafluoro-λ⁵-phosphanuide
Synonyms: ethyl cyano({[(dimethyliminio)(morpholin-4-yl)methoxy]imino})formate hexafluorophosphate
Molecular Weight (g/mol): 428.27
Molecular Formula: C12H19F6N4O4P
InChi Key: GPDHNZNLPKYHCN-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NOC(N1CCOCC1)=[N+](C)C)C#N
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)