missing translation for 'onlineSavingsMsg'
Learn More

COMU™, 97%

Product Code. 11326435 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11326435 5 g Glass bottle 5g
11336435 10 g Glass bottle 10g
11346435 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326435 Supplier Thermo Scientific Acros Supplier No. 439480050
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Coupling reagent | CAS: 1075198-30-9 | C12H19F6N4O4P | 428.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075198-30-9
Molecular Formula C12H19F6N4O4P
Molecular Weight (g/mol) 428.27
MDL Number MFCD11975052 MFCD24368564 MFCD11975052
InChI Key GPDHNZNLPKYHCN-UHFFFAOYSA-N
Synonym comu, 1-cyano-2-ethoxy-2-oxoethylidenaminooxy dim, 1-cyano-2-ethoxy-2-oxoethylidenaminooxy dimethylamino-morpholinocarbenium hexafluorophosphate, 1-cyano-2-ethoxy-2-oxoethylideneaminooxy-dimethylamino-morpholino-carbenium hexafluorophosphate, z-1-cyano-2-ethoxy-2-oxoethylidene amino oxy-n,n-dimethyl morpholin-4-yl methaniminium hexafluorophosphate, 4-e-1-cyano-2-ethoxy-2-oxoethylidene amino oxidanyl dimethylamino methylidene-4??-morpholin-4-ylium hexafluorophosphate
PubChem CID 44471148
IUPAC Name [[(Z)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-morpholin-4-ylmethylidene]-dimethylazanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NOC(N1CCOCC1)=[N+](C)C)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 151.4°C to 152.9°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 428.27
Percent Purity 97%
Physical Form Powder or Solid
Chemical Name or Material COMU*
Show More Show Less
compliance-icons
Product Identifier
  • (1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholino-carbenium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.