accessibility menu, dialog, popup

UserName

H3COCH3OCH3O

CAS RN 108-65-6

IUPAC Name: 1-methoxypropan-2-yl acetate
Synonyms: methoxyisopropyl acetate propylene glycol methyl ether acetate propylene glycol ME ether acetate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: LLHKCFNBLRBOGN-UHFFFAOYNA-N
SMILES: COCC(C)OC(C)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)