accessibility menu, dialog, popup

UserName

CAS RN 108-80-5

ONHONHOHN

CAS RN 108-80-5

IUPAC Name: 1,3,5-triazinane-2,4,6-trione
Synonyms: isocyanuric acid isozyanursaeure isocyanursaeure
Molecular Weight (g/mol): 129.08
Molecular Formula: C3H3N3O3
InChi Key: ZFSLODLOARCGLH-UHFFFAOYSA-N
SMILES: O=C1NC(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)