missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric acid, 98%

Product Code. 10635822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10635822 250 g Plastic bottle 250g
10184340 25 g Plastic bottle 25g
10701081 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10635822 Supplier Thermo Scientific Acros Supplier No. 181022500
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1

Specifications

Color White to Beige
Quantity 250 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder or Lumps
Assay Percent Range 98%
Beilstein 26, 239
Merck Index 15, 2690
Formula Weight 129.07
Percent Purity ≥97.5%
Chemical Name or Material Cyanuric acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.