accessibility menu, dialog, popup

UserName

CH3OOH3C

CAS RN 109-87-5

IUPAC Name: dimethoxymethane
Synonyms: dimethoxymethane methylene glycol dimethylether methylal formaldehyde methyl ketal formaldehyde dimethylacetal dimethyl formal anesthenyl formal metylal methylene dimethyl ether
Molecular Weight (g/mol): 76.1
Molecular Formula: C3H8O2
InChi Key: NKDDWNXOKDWJAK-UHFFFAOYSA-N
SMILES: COCOC
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (6)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)