accessibility menu, dialog, popup

UserName

CAS RN 110-72-5

H3CNH2NH3

CAS RN 110-72-5

IUPAC Name: (2-azaniumylethyl)(ethyl)azanium
Synonyms: (2-ammonioethyl)(ethyl)azanium
Molecular Weight (g/mol): 90.17
Molecular Formula: C4H14N2
InChi Key: SCZVXVGZMZRGRU-UHFFFAOYSA-P
SMILES: CC[NH2+]CC[NH3+]
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)