missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylethylenediamine, TRC

Product Code. 30384098
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30384098 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30384098

Brand: Toronto Research Chemicals TRCE9178131G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethylethylenediamine
CAS 110-72-5
Molecular Formula C4H12N2
Quantity 1 g
Synonym N-Ethyl-1,2-ethanediamine, N-Ethyl-1,2-diaminoethane, N-(2-Aminoethyl)ethylamine, 2-(Ethylamino)ethylamine, 2-(Ethylamino)ethanamine, 1-Ethylethylenediamine, N1-Ethyl-1,2-ethanediamine
InChI Formula InChI=1S/C4H12N2/c1-2-6-4-3-5/h6H,2-5H2,1H3
SMILES CCNCCN
IUPAC Name N'-ethylethane-1,2-diamine
Molecular Weight (g/mol) 88.15
Formula Weight 88.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.