accessibility menu, dialog, popup

UserName

OHN

CAS RN 111060-52-7

IUPAC Name: 2-(dibenzylamino)-3-phenylpropan-1-ol
Synonyms: 2-(dibenzylamino)-3-phenylpropan-1-ol
Molecular Weight (g/mol): 331.46
Molecular Formula: C23H25NO
InChi Key: ZXNVOFMPUPOZDF-UHFFFAOYNA-N
SMILES: OCC(CC1=CC=CC=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results