accessibility menu, dialog, popup

UserName

NH2(S)HOOOHO

CAS RN 1118-90-7

IUPAC Name: (2S)-2-aminohexanedioic acid
Synonyms: aminoadipate acid, α-aminoadipic acid, 2-aminohexanedioic acid, 2-aminoadipic acid, 2 aminoadipic
Molecular Weight (g/mol): 161.16
Molecular Formula: C6H11NO4
InChi Key: OYIFNHCXNCRBQI-BYPYZUCNSA-N
SMILES: N[C@@H](CCCC(O)=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)