missing translation for 'onlineSavingsMsg'
Learn More

L-2-Aminoadipic acid, 98%, Thermo Scientific™

Product Code. 10731355 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10731355 5 g Glass bottle 5g
10399250 250 mg Glass bottle 250mg
10422855 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10731355 Supplier Thermo Scientific Supplier No. 302920050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-90-7
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00002636
InChI Key OYIFNHCXNCRBQI-BYPYZUCNSA-N
Synonym l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
PubChem CID 92136
ChEBI CHEBI:37023
IUPAC Name (2S)-2-aminohexanedioic acid
SMILES N[C@@H](CCCC(O)=O)C(O)=O

Specifications

Melting Point 206°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2C(CH2)3CH(NH2)CO2H
Quantity 5 g
Specific Rotation Condition +22° (25°C c=2,5 NHCl)
Specific Rotation + 22
Formula Weight 161.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-2-Aminoadipic acid, 98%
Show More Show Less

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.