accessibility menu, dialog, popup

UserName

OH(S)H2N

CAS RN 112068-01-6

IUPAC Name: (2S)-2-(hydroxydiphenylmethyl)pyrrolidin-1-ium
Synonyms: (2S)-2-(hydroxydiphenylmethyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 254.35
Molecular Formula: C17H20NO
InChi Key: OGCGXUGBDJGFFY-INIZCTEOSA-O
SMILES: OC([C@@H]1CCC[NH2+]1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)