accessibility menu, dialog, popup

UserName

CAS RN 112108-73-3

CH3ClFNOO

CAS RN 112108-73-3

IUPAC Name: 1-chloro-5-fluoro-2-methyl-4-nitrobenzene
Synonyms: 1-chloro-5-fluoro-2-methyl-4-nitrobenzene
Molecular Weight (g/mol): 189.57
Molecular Formula: C7H5ClFNO2
InChi Key: YXVJHZWHPLOEAP-UHFFFAOYSA-N
SMILES: CC1=CC(=C(F)C=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results