accessibility menu, dialog, popup

UserName

FBrFNH2

CAS RN 112279-60-4

IUPAC Name: 4-bromo-2,5-difluoroaniline
Synonyms: 4-bromo-2,5-difluoroaniline
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: XOYHFIQPPOJMFK-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results