accessibility menu, dialog, popup

UserName

FSN

CAS RN 1123-98-4

IUPAC Name: 2-fluoro-1,3-benzothiazole
Synonyms: 2-fluoro-1,3-benzothiazole
Molecular Weight (g/mol): 153.17
Molecular Formula: C7H4FNS
InChi Key: QVWCHVAUHZEAAT-UHFFFAOYSA-N
SMILES: FC1=NC2=CC=CC=C2S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results