accessibility menu, dialog, popup

UserName

CAS RN 1126-79-0

H3CO

CAS RN 1126-79-0

IUPAC Name: butoxybenzene
Synonyms: butyl phenyl ether
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: YFNONBGXNFCTMM-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results