accessibility menu, dialog, popup

UserName

NH2

CAS RN 1135-12-2

IUPAC Name: 4-benzylaniline
Synonyms: 4-benzylaniline
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: WDTRNCFZFQIWLM-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results