accessibility menu, dialog, popup

UserName

CAS RN 114045-96-4

OOOHF

CAS RN 114045-96-4

IUPAC Name: 4-(benzyloxy)-2-fluorobenzoic acid
Synonyms: 4-(benzyloxy)-2-fluorobenzoic acid
Molecular Weight (g/mol): 246.24
Molecular Formula: C14H11FO3
InChi Key: HWQRXVRKZFDKLJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results